Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drebrin 1, Mouse anti-Human, DyLight 488, Clone: DBN1/2879, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279901G

Catalog No. NB012177


Add to cart

Description

Description

Drebrin 1 Monoclonal specifically detects Drebrin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Drebrin 1
Monoclonal
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
DBN1
Recombinant fragment (aa 150-281) of human Drebrin 1 protein (exact sequence is proprietary)
0.1 mL
Neuroscience
Human
IgG1 κ
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
DBN1/2879
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
D0S117E, Developmentally-regulated brain protein, DKFZp434D064, drebrin, drebrin 1, drebrin E, drebrin E2, drebrin-1
Mouse
Bioreactor concentrate
RUO
Primary
Store at 4°C in the dark.
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit