Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Drebrin 1, Mouse anti-Human, DyLight 650, Clone: DBN1/2879, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP279901C
Description
Drebrin 1 Monoclonal specifically detects Drebrin 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Drebrin 1 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DBN1 | |
Recombinant fragment (aa 150-281) of human Drebrin 1 protein (exact sequence is proprietary) (Uniprot: Q16643) | |
0.1 mL | |
Neuroscience | |
Human | |
IgG1 κ |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DBN1/2879 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
D0S117E, Developmentally-regulated brain protein, DKFZp434D064, drebrin, drebrin 1, drebrin E, drebrin E2, drebrin-1 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title