Learn More
Enzo Life SciencesSupplier Diversity Partner Drebrin monoclonal antibody (M2F6) (100 µg)  DFS Item
Manufacturer:  Enzo Life SciencesSupplier Diversity Partner ADINBA110E
Clone: M2F6. Host: Mouse. Isotype: IgG1. Immunogen: Chicken Drebrin E. UniProt: P18302. Species Reactivity: Human, Mouse, Rat, Rabbit, Chicken, Cat, Bovine. Applications: ICC, IHC, IP, WB. Recommended dilutions: Immunohistochemistry (10µg/ml). Western Blot (1µg/ml). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein A affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol.
Catalog No. 50-666-986