Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DREF Mouse anti-Human, Clone: PCRP-ZBED1-1E1, Novus Biologicals™
Mouse Monoclonal Antibody
$265.00
Specifications
Antigen | DREF |
---|---|
Clone | PCRP-ZBED1-1E1 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array |
Applications | Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB122546
|
Novus Biologicals
NBP30753620UG |
20 μg |
Each of 1 for $265.00
|
|
|||||
Description
DREF Monoclonal specifically detects DREF in Human samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
DREF | |
0.2 mg/ml | |
Western Blot, Flow Cytometry, Immunofluorescence, Peptide Array | |
Unconjugated | |
Mouse | |
Human | |
Ac-like transposable element, ALTEDNA replication-related element binding factor, hDREF, KIAA0785dREF homolog, Putative Ac-like transposable element, putative Ac-like transposon, TRAMPDREFBED-type zinc finger domain-containing protein 1, zinc finger BED domain-containing protein 1, zinc finger, BED-type containing 1 | |
Recombinant full-length human DREF protein | |
Primary | |
Store at 4°C. |
PCRP-ZBED1-1E1 | |
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
9189 | |
IgG2a | |
Protein A or G purified |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title