Learn More
Sklar InstrumentsSupplier Diversity Partner Dressing Jar, Sklar, Stainless Steel. Sklar Dressing Jars are ideal for storing medical supplies such as tongue depressors, cotton balls, and cotton swabs. This jar measures 6-3/4 in. x 7-3/4 in. and has a capacity of 4-3/4 quarts. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 101543
Dressing Jar, Sklar, Stainless Steel. Sklar Dressing Jars are ideal for storing medical supplies such as tongue depressors, cotton balls, and cotton swabs. This jar measures 6-3/4 in. x 7-3/4 in. and has a capacity of 4-3/4 quarts. It also features a slip over lid and a recessed handle.
Catalog No. 50-143-7051