Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRGX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | DRGX |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP191316
|
Novus Biologicals
NBP191316 |
100 μL |
Each of 1 for $482.50
|
|
|||||
Description
DRGX Polyclonal specifically detects DRGX in Human samples. It is validated for Western Blot.Specifications
DRGX | |
Polyclonal | |
Rabbit | |
NP_001073989 | |
644168 | |
Synthetic peptide directed towards the N terminal of human DRGX. Peptide sequence MVLMSCDQRLISLLLVGTATFGNHSSGDFDDGFLRRKQRRNRTTFTLQQL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
dorsal root ganglia homeobox, DRG11, paired related homeobox-like 1, paired-like homeodomain trancription factor DRG11, paired-related homeobox protein-like 1, PRRXL1 | |
DRGX | |
IgG |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title