Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRGX Rabbit anti-Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310397100UL
Description
DRGX Polyclonal specifically detects DRGX in Rat samples. It is validated for Western Blot.Specifications
DRGX | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
dorsal root ganglia homeobox, DRG11, paired related homeobox-like 1, paired-like homeodomain trancription factor DRG11, paired-related homeobox protein-like 1, PRRXL1 | |
The immunogen is a synthetic peptide directed towards the middle region of Rat DRGX (NP_665710). Peptide sequence GAKEPMAEVTPPPVRNINSPPPGDQARGKKEALEAQQSLGRTVGPAGPFF | |
100 μg | |
Primary | |
Rat | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
644168 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction