Learn More
Thermo Scientific™ DRI™ Barbiturate Drugs of Abuse Assays
Achieve high analytical accuracy and eliminate time-consuming steps in reagent preparation with ready-to-use assays in liquid form.
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Control Sets | MGC Multi-Drug Controls |
Detectable Analytes | Secobarbital |
DoA Calibrators | DRI Multi-drug Calibrators |
Type | Reagent |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-278-313
|
Thermo Scientific™
0225 |
1 x 100 mL |
Each for $2,419.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-357
|
Thermo Scientific™
10015648 |
3 x 18 mL |
Each for $1,251.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Achieve high analytical accuracy and eliminate time-consuming steps in reagent preparation with ready-to-use assays in liquid form. Thermo Scientific™ DRI™ Barbiturate Drugs of Abuse Assays have been field-tested on a wide range of analyzers, and they are enthusiastically accepted in laboratories where cost-control and optimal productivity are key issues.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Variety of calibrators and multiconstituent control sets
Specifications
2°C to 8°C | |
Secobarbital | |
Reagent |
MGC Multi-Drug Controls | |
DRI Multi-drug Calibrators |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.