Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ DRI™ Fentanyl II Drugs of Abuse Calibrators

Catalog No. p-9413004
Click to view available options
Description:
DRI Fentanyl II Cutoff Calibrator 
DRI Fentanyl II Negative Calibrator
Concentration:
0 ng/mL 
1 ng/mL 
Detectable Analytes:
Fentanyl
Negative Calibrator
This item is not returnable. View return policy
This item is not returnable. View return policy

Ensure accurate calibration with the Thermo Scientific™ DRI™ Fentanyl II Drugs of Abuse Calibrators, designed for urine screening with the Thermo Scientific DRI Fentanyl II Drugs of Abuse Assay.

Ensure accurate calibration with the Thermo Scientific DRI Fentanyl II Drugs of Abuse Calibrators, designed for urine screening with the Thermo Scientific DRI Fentanyl II Drug of Abuse Assay. The calibrators, sold separately, include a negative urine calibrator and a cutoff calibrator spiked with 1 ng/mL of fentanyl.

  • Specifically detect fentanyl and its major metabolite, norfentanyl, at physiologically relevant concentrations in human urine
  • Low cross-reactivity to other non-fentanyl synthetic opioids and over-the-counter medication 
  • High sensitivity with a cutoff concentration of 1 ng/mL 
  • Superior assay precision
  • Excellent correlation to LC-MS/MS
  • Liquid, Ready-to-Use 
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Description DRI Fentanyl II Cutoff Calibrator 
Concentration 1 ng/mL 
Detectable Analytes Fentanyl
Sample Type Urine
Packaging 2 Bottles
Quantity 2 x 10 mL 

For sale in the U.S.A. and Canada only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.