Learn More
Thermo Scientific™ DRI™ Methadone Drugs of Abuse Assays
Use Thermo Scientific™ DRI™ Methadone Drugs of Abuse Assays to achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation.
Specifications
Content And Storage | 2°C to 8°C |
---|---|
Control Sets | MGC Multi-Drug Controls |
Detectable Analytes | Methadone |
DoA Calibrators | DRI Multi-drug Calibrators |
Sample Type | Urine |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
23-278-325
|
Thermo Scientific™
0596 |
1 x 100 mL |
Each for $2,442.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-278-375
|
Thermo Scientific™
10016403 |
3 x 18 mL |
Each for $724.00
|
N/A | |||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
Use Thermo Scientific™ DRI™ Methadone Drugs of Abuse Assays to achieve high analytical accuracy while eliminating time-consuming steps in reagent preparation. These liquid, ready-to-use assays offer applications for an array of clinical chemistry analyzers. Field-tested on a wide range of analyzers, these assays are enthusiastically accepted in laboratories where cost control and optimal productivity are key issues.
- Qualitative or semi-quantitative options for results
- Excellent correlation to GC/MS
- Variety of calibrators and multiconstituent control sets
Specifications
2°C to 8°C | |
Methadone | |
Urine |
MGC Multi-Drug Controls | |
DRI Multi-drug Calibrators | |
Reagent |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.