Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dried Aluminum Hydroxide Gel, Powder, USP, 76.5%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Pail
21645-51-2
AlH6O3
81.03
MXRIRQGCELJRSN-UHFFFAOYSA-N
alumanetriol
O.O.O.[Al]
21645-51-2
AlH6O3
81.03
MXRIRQGCELJRSN-UHFFFAOYSA-N
alumanetriol
O.O.O.[Al]

AL226, 21645-51-2, Al(OH)3

Spectrum™ Dried Aluminum Hydroxide Gel, Powder, USP is used as an antacid. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 8 ppm
Percent Purity 76.5%
Chloride 0.0085
Packaging Poly Pail
pH Range 10
Sulfate 0.006
Quantity 2.5 kg