Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dried Ferrous Sulfate, Powder, USP, 86-89%, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
13463-43-9
FeH2O5S
169.92
XBDUTCVQJHJTQZ-UHFFFAOYSA-L
λ²-iron(2+) hydrate sulfate
O.[Fe++].[O-]S([O-])(=O)=O
13463-43-9
FeH2O5S
169.92
XBDUTCVQJHJTQZ-UHFFFAOYSA-L
λ²-iron(2+) hydrate sulfate
O.[Fe++].[O-]S([O-])(=O)=O

F1065, 13463-43-9, FeSO4•xH2O

Spectrum™ Dried Ferrous Sulfate, Powder, USP is used to prevent or treat low levels of iron in the blood. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Arsenic (As) 3 ppm
Percent Purity 86 to 89%
Insoluble Matter 0.0005
Lead (Pb) 10 ppm
Mercury (Hg) 3 ppm
Packaging Poly Pail
Quantity 2.5 kg