Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRIERITE™, Indicating, 8 Mesh, J.T. Baker™
Click to view available options
Quantity:
454 g
2.3 kg
Specifications
Specifications
Quantity | 454 g |
Mesh Size | 8 Mesh |
Specific Gravity | 1L = 1.87kg |
Chemical Name or Material | DRIERITE™, Indicating |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction