Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Drierite™, with indicator, 10-20 mesh, ACROS Organics™

Click to view available options
Quantity:
500g
2kg
10kg
Packaging:
Glass bottle
Plastic Drum
7778-18-9,7646-79-9
CaO4S
136.134
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
24497
CHEBI:31346
calcium;sulfate
[O-]S(=O)(=O)[O-].[Ca+2]
7778-18-9,7646-79-9
CaO4S
136.134
OSGAYBCDTDRGGQ-UHFFFAOYSA-L
24497
CHEBI:31346
calcium;sulfate
[O-]S(=O)(=O)[O-].[Ca+2]
Adsorbent, Indicator
Melting Point 1450.0°C
Cobalt Chloride 3.25%
Calcium Oxide 39.09%
Carbon Dioxide 0.68%
Color Blue
Quantity 2kg
UN Number 3077
Packaging Glass bottle
SiO2 and Insolubles SiO2 and insolubles: 0.58%
R2O 0.29%
Solubility Information Solubility in water: slightly soluble. Other solubilities: slightly soluble in glycerol, practically insoluble in most common organic, solvents
Sodium Chloride 0.23%
Sulfite 55.82%
Formula Weight 136.13
Physical Form Irregular Granules
Chemical Name or Material Drierite™
Show More Show Less

GHS H Statement
May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. Suspected of causing genetic defects. May cause cancer by inhalation. May damage fertility. Causes serious eye irritation. Toxic to aquatic life with long lasting effects.

GHS P Statement
Obtain special instructions before use. IF ON SKIN: Wash with plenty of soap and water. IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. IF exposed or concerned: Get medical advice/attention.

Danger