Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Drierite™, without indicator, 4 mesh, ACROS Organics™
Click to view available options
Quantity:
500g
2kg
10kg
Packaging:
Glass bottle
Plastic drum
Description
Adsorbent, Indicator
Specifications
Melting Point | 1450.0°C |
Composition | Typical chemical analysis: CaO: 40.80%, SO3: 58.80%, CO2: 0.60%, R2O: 0.14%, SiO2 and insolubles: 0.15%, NaCl: 0.18% |
Quantity | 500g |
Packaging | Glass bottle |
Solubility Information | Solubility in water: ca. 0.25g/L (20°C). Other solubilities: insoluble in organic liquids, slightly soluble in glycerol |
Formula Weight | 136.14 |
Physical Form | Granular |
Chemical Name or Material | Drierite™ |