DRIFIRE™ Fortrex™ Two-Piece Flight Suits Available on GSA/VA Contract for Federal Government customers only.

Unique design enables reduced binding, increased range of motion and reduced snag hazards

Manufacturer: DRIFIRESupplier Diversity Partner PHX2550FSP3X MC

 View more versions of this product

Catalog No. 17-000-260

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Specifications

    Product Type Pant
    Certifications/Compliance A2CU
    Material Fabric
    Color Multicam
    Size 3X-Large
    For Use With (Application) US army, US coast guard, air force, US law enforcement/defense contractors and US SOCOM
    Resistance Flame
    Closure Type Velcro™ and zipper

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit