Learn More
DRIFIRE™ Heavyweight Long Sleeve Shirts
SDP
Catalog Number 17000184
Click to view available options
Color:
Black
Coyote Brown
Desert Sand
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy
Can be wear alone or as part of layering system for increased protection and warmth
  • Made of soft and comfortable, DRIFIRE fabric that prevents chafing
  • Suitable for cold climates
  • Flame resistant
  • Dropped-shoulder, flat-lock seams for no-rub comfort
  • Controls odor
  • Moisture-wicking and dries four times faster than cotton
Size Medium
Material Heavyweight; Flame resistant
Color Black