Learn More
DRIFIRE™ Heavyweight Long Sleeve Shirts
Click to view available options
Color:
Black
Coyote Brown
Desert Sand
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Made of soft and comfortable, DRIFIRE fabric that prevents chafing
- Suitable for cold climates
- Flame resistant
- Dropped-shoulder, flat-lock seams for no-rub comfort
- Controls odor
- Moisture-wicking and dries four times faster than cotton
Specifications
Size | Medium |
Material | Heavyweight; Flame resistant |
Color | Desert Sand |