Learn More
Dunlop™ Onguard™ Chest Waders with Cleated Outsole
Superior waterproof protection that resists oil, gas, grease, and saltwater in every condition.
Supplier: Dunlop™ 86067.14
This item is not returnable.
View return policy
Description
These durable chest waders are made of heavy duty PVC and boast cleated outsoles for excellent flexibility and traction clogging prevention, even in wet or muddy terrain.
- Chest high, with PVC upper and non-absorbent polyester backing
- Injection molded, seamless boot construction
- Steel midsole and adjustable polyester and nylon strap
- Excellent cold weather flexibility
- Can be folded or rolled without cracking
- Top drawstring closure for snug fit
- Non-corrosive, reinforced suspender buttons
- Built-in belt loops with adjustable belt
- Replaceable cushion insoles
- Made in the USA
Specifications
ASTM F2413-11 M I/75 C/75 SD | |
Black | |
PVC | |
14 | |
Polyester/Nylon |
Drawstring | |
Utilities, Municipalities, Aquaculture, and Commercial Fishing | |
Seamless | |
Cleated Outsole, Steel Midsole, Replaceable Cushion Insole | |
Steel |