DRIFIRE™ Lightweight Military Boxer Briefs

Used as base layer for other DriFire garments

Manufacturer: DRIFIRESupplier Diversity Partner DF4505BBMD DS

 View more versions of this product

Catalog No. 17-000-478

  Check Availability
Add to Cart

    Specifications

    Size (Metric) Waist 78.7cm
    Size Medium
    Size (English) Waist 31 in.
    Color Desert Sand

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit