DRIFIRE™ Lightweight Military Boxer Briefs Available on GSA/VA Contract for Federal Government customers only.

Used as base layer for other DriFire garments

Manufacturer: DRIFIRESupplier Diversity Partner DF2446BB 2X NB

 View more versions of this product

Catalog No. 17-000-395

  Check Availability
Add to Cart

    Specifications

    Material Fabric
    Size (Metric) Waist 101.6cm
    Size (English) Waist 40 in.
    For Use With (Application) Other DRIFIRE garments, beginning for any uniform
    Size 2XL
    Color Navy Blue

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit