Learn More
DRIFIRE™ Lightweight Short-Sleeve Tees
Click to view available options
Color:
Black
Brown
Desert Sand
Green
Navy
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Made of soft fabric to prevent chafing
- With ribbed collar, ideal base layer and no rub seams
- Controls odor
- Moisture-wicking and fast-drying
- Permanent flame resistance HRC 1
- Available in Black, Desert Sand, Navy Blue
- Sizes available: Small to 3X-Large
Specifications
Size | Large |
Material | Soft, Lightweight Fabric |
Color | Desert Sand |