Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DRIFIRE™ Navair One-Piece Flight Suits
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- Made of proprietary flame resistant material with 27-P design
- Durable, cool and breathable fabric that minimizes heat buildup
- With thigh-vents to cool femoral artery
- Moisture wicking and fast-drying
- High tensile and tear strengths
- Navy and USMC “Safe to Fly”
- Meets NAVAIR specifications
- Available in khaki color with different sizes ranging from Small to XXX-Large
US Navy, US Coast Guard, US Marines, US Law Enforcement/ Defense Contractors and MRO (Maintence Repair Operations)
Specifications
Size | 2X-Large |
Material | Fabric |
Color | Khaki |
Certifications/Compliance | NAVAIR |
For Use With (Application) | US navy, US coast guard, US marines, US law enforcement/ defense contractors and MRO |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction