Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRIFIRE™ Navair One-Piece Flight Suits

With adjustable side tabs for a perfect fit around the torso

$650.50 - $716.00

Specifications

Certifications/Compliance NAVAIR
Color Khaki
Material Fabric
For Use With (Application) US navy, US coast guard, US marines, US law enforcement/ defense contractors and MRO
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
17-000-268
View Documents
DRIFIRE™
PHX17129450TNSMR
Small
Each for $650.50
Add to cart
 
17-000-270
View Documents
DRIFIRE™
PHX17129450TNLGR
Large
Each for $650.50
Add to cart
 
17-000-272
View Documents
DRIFIRE™
PHX17129450TN2XR
2X-Large
Each for $716.00
Add to cart
 
17-000-337
View Documents
DRIFIRE™
PHX17129450TNXLR
X-Large
Each for $650.50
Add to cart
 
17-000-335
View Documents
DRIFIRE™
PHX17129450TNMDR
Medium
Each for $650.50
Add to cart
 
17-000-339
View Documents
DRIFIRE™
PHX17129450TN3XR
3X-Large
Each for $716.00
Add to cart
 
Description

Description

  • Made of proprietary flame resistant material with 27-P design
  • Durable, cool and breathable fabric that minimizes heat buildup
  • With thigh-vents to cool femoral artery
  • Moisture wicking and fast-drying
  • High tensile and tear strengths
  • Navy and USMC “Safe to Fly”
  • Meets NAVAIR specifications
  • Available in khaki color with different sizes ranging from Small to XXX-Large

US Navy, US Coast Guard, US Marines, US Law Enforcement/ Defense Contractors and MRO (Maintence Repair Operations)

Specifications

Specifications

NAVAIR
Khaki
Fabric
US navy, US coast guard, US marines, US law enforcement/ defense contractors and MRO
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit