Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DRIFIRE™ Two-Piece Navy Flight Suits

Designed for Naval and Marine Aviators with end-user input

$621.00 - $683.50

Specifications

Color Khaki
Description Modacrylic, Cotton, Para Aramid™, Anti-stat
Material Modacrylic, Cotton
For Use With (Application) Naval and marine aviators with end-user input
Resistance Flame
View More Specs

Products 7
Catalog Number Mfr. No. Closure Type Size Price Quantity & Availability  
Catalog Number Mfr. No. Closure Type Size Price Quantity & Availability  
17-000-276
View Documents Promotion Details
DRIFIRE™
PHX27129JA450TNMDR
Zipper Medium
Each for $621.00
Add to cart
 
17-000-278
View Documents Promotion Details
DRIFIRE™
PHX27129JA450TNXLR
Zipper X-Large
Each for $621.00
Add to cart
 
17-000-279
View Documents Promotion Details
DRIFIRE™
PHX27129JA450TN2XR
Zipper 2X-Large
Each for $683.50
Add to cart
 
17-000-340
View Documents Promotion Details
DRIFIRE™
PHX27129JA450TNXSR
Zipper X-Small
Each for $621.00
Add to cart
 
17-000-341
View Documents Promotion Details
DRIFIRE™
PHX27129JA450TNSMR
Zipper Small
Each for $621.00
Add to cart
 
17-000-343
View Documents Promotion Details
DRIFIRE™
PHX27129JA450TNLGR
Zipper Large
Each for $621.00
Add to cart
 
17-000-346
View Documents Promotion Details
DRIFIRE™
PHX27129JA450TN3XR
Zipper 3X-Large
Each for $683.50
Add to cart
 
Description

Description

  • Permanent, inherent flame resistance that won't wash or wear out
  • Controls odor
  • Increased durability and FR performance
  • Unique design features to improve fit, increase range of motion and reduce snag hazards
  • Navy and USMC “Safe to Fly”
  • Meets NAVAIR specifications
  • Available in khaki color in different sizes ranging from X-Small to XXX-Large

Two-Piece Navy Flight Suit Jackets:

  • Angled shoulder pockets for easy access
  • Adjustable waist tabs for customized torso fit

US Navy, US Coast Guard, US Marines and MRO (Maintence Repair Operations)

Specifications

Specifications

Khaki
Modacrylic, Cotton
Flame
Modacrylic, Cotton, Para Aramid™, Anti-stat
Naval and marine aviators with end-user input
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit