Learn More
Cole-Parmer Masterflex I/P Digital Modular Drive with Remote Input/Output, 6 to 650 rpm; 115/230 VAC
Manufacturer:  Cole-Parmer 0759120
Encompass_Preferred
I/P modular drives feature reversible pumping while allowing you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering and critical transfer applications. Splash-resistant ABS plastic controller housing and epoxy finished steel drive housing are IP33 rated. A 6-ft (1.8-m) cable connects drive and controller—optional extension cable 07557-51 is available to extend distance to 15 ft (4.5 m).
Catalog No. NC1736692