Learn More
Cole-Parmer Masterflex L/S Digital Precision Modular Drive with Remote I/O and Benchtop Controller, 6 to 600 rpm; 90 to 260 VAC
Manufacturer:  Cole-Parmer 0755700
Encompass_Preferred
L/S modular drives feature reversible pumping and allow you the flexibility to separate the motor from the controller. These accurate modular drives have powerful motors, speed display, and ±0.25% speed control for precise flow regulation—ideal for metering applications.
Catalog No. NC1453998