Learn More
Unchained LabsSupplier Diversity Partner Big Lunatic Calibration Plate DFS Item
Manufacturer:  Unchained LabsSupplier Diversity Partner 7012013
Customer calibration plate for wavelength, positon and pump calibration. For use with Big Lunatic and DropSense96. (Previously product code 4-30201)
Catalog No. 50-112--5941