Learn More
Medchemexpress LLC HY-B1032 500mg Medchemexpress, Dropropizine CAS:17692-31-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1032500MG
Encompass_Preferred
Medchemexpress, HY-B1032 500mg Dropropizine CAS:17692-31-8 Dropropizine ((±)-Dropropizine) is a peripheral antitussive agent that acts by inhibiting cough reflex through its action on the peripheral receptors and their afferent conductors. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2681
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.