Learn More
Genetex IncSupplier Diversity Partner Anti-DRP2 antibody [N3C1], Internal DFS Item
Manufacturer:  Genetex IncSupplier Diversity Partner GTX112512
The Anti-DRP2 antibody [N3C1], Internal from GeneTex is a Rabbit Polyclonal antibody to DRP2. The Anti-DRP2 antibody [N3C1], Internal Recognizes Mouse. The Anti-DRP2 antibody [N3C1], Internal has been validated for the following Applications: WB.
Catalog No. NC1471422