Drucker™ Accessories for SampleSafe™ Storage Lock Box
Optional accessories for use with SampleSafe Storage Lock Box
Manufacturer: Drucker Diagnostics 03100120039
 View more versions of this product
Catalog No. 22-029-580
  Check Availability
Add to Cart
    Specifications
    Item Description Rigid plastic plate used to separate samples from the cold packs  preventing unwanted ice pack contact with sensitive samples.
    For Use With (Equipment) Drucker SampleSafe LB1 and LB10 Lock Boxes
    Product Type False Bottom
  • Description & Specifications
Related Products
Product Content Feedback