Drummond Scientific Calibrated Micropipets Available on GSA/VA Contract for Federal Government customers only.

Anticoagulant treatment for clinical applications

Manufacturer: Drummond Scientific 2000075

 View more versions of this product

Catalog No. 21-180-17

  Check Availability
Add to Cart

    Specifications

    Includes 250 pipets and one 15 in. aspirator tube
    Accuracy ±0.5%
    Calibrated Yes
    Color Purple
    For Use With (Application) Anticoagulant treatment for clinical applications, ideal for liquid transfer applications
    Capacity (Metric) 75μL
    Material Glass
    Product Type Calibrated Micropipet

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit