Learn More
Drummond Scientific Original Pipet-Aid™ Pipet Controller Available on GSA/VA Contract for Federal Government customers only.
Safe and easy-to-operate alternative to mouth pipetting in the laboratory
Manufacturer:  Drummond Scientific 4000110
 View more versions of this product
Catalog No. 13-681-15
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
Glass or plastic pipets
EZ Grip TC Filter Nosepiece
Standard
1 to 150mL
Pipetting of biohazardous materials
60Hz
110V
Filler/Dispenser
Accessories