Drummond Scientific Original Pipet-Aid™ Pipet Controller Available on GSA/VA Contract for Federal Government customers only.

Safe and easy-to-operate alternative to mouth pipetting in the laboratory

Manufacturer: Drummond Scientific 4000111TC

 View more versions of this product

Catalog No. 13-681-15D

  Check Availability
Add to Cart

    Specifications

    Autoclavable Yes
    For Use With (Application) Pipetting of biohazardous materials
    For Use With (Equipment) Glass or plastic pipets
    Hertz 60Hz
    Tip Style EZ Grip TC Filter Nosepiece
    Voltage 110V
    Model Dual Pump Filtration Unit Complete†,Dual Pump Filtration Unit Complete†
    Product Type Filler/Dispenser
    Volume (Metric) Range 1 to 150mL
    Volume (Metric) 1 to 150mL

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit