Drummond Scientific Original Pipet-Aid™ Pipet Controller

Safe and easy-to-operate alternative to mouth pipetting in the laboratory

Manufacturer: Drummond Scientific 4000221

 View more versions of this product

Catalog No. 13-681-16A

  Check Availability

This item is a non-standard voltage; please ensure you are ordering the correct product.

Please contact customer service for assistance: 1-800-766-7000.

Add to Cart

    Specifications

    Autoclavable Yes
    For Use With (Application) Pipetting of biohazardous materials
    For Use With (Equipment) Glass or plastic pipets
    Tip Style EZ Grip TC Filter Nosepiece
    Voltage 220V
    Model Complete†,Complete†
    Product Type Filler/Dispenser
    Volume (Metric) Range 1 to 150mL
    Volume (Metric) 1 to 150mL

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit