Drummond Scientific Replacement Bores for Drummond Micropipetters Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: Drummond Scientific 3000275G

 View more versions of this product

Catalog No. 21-169F

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) 100/200μL Drummond™ micropipetter, digital microdispenser
    Material Glass
    Color Blue,Blue
    Product Type Bore

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit