Learn More
Drummond Scientific Wiretrol™ I Calibrated Micropipets Available on GSA/VA Contract for Federal Government customers only.
Easy-to-read volume line won't wash off
Manufacturer:  Drummond Scientific 50001005
 View more versions of this product
Catalog No. 21-175A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
To contain
For a wide variety of applications, Wiretrol™ will accept both high and low viscosity liquids including corrosives
5μL
Calibrated Micropipet
White
Handy dispenser cap, handle, Stainless Steel plunger
Glass