Drummond Scientific Wiretrol™ I Calibrated Micropipets Available on GSA/VA Contract for Federal Government customers only.

Easy-to-read volume line won't wash off

Manufacturer: Drummond Scientific 50001100

 View more versions of this product

Catalog No. 21-175F

  Check Availability
Add to Cart

    Specifications

    Calibration To contain
    Color Blue
    For Use With (Application) For a wide variety of applications, Wiretrol™ will accept both high and low viscosity liquids including corrosives
    Includes Handy dispenser cap, handle, Stainless Steel plunger
    Capacity (Metric) 100μL
    Material Glass
    Product Type Calibrated Micropipet

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit