Learn More
Thermo Scientific™ Drums for Digital Cel-Gro Tissue Culture Rotators
Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V
$216.00 - $433.00
Specifications
Product Type Rotating Drum
View More Specs
Products 9
Catalog Number Mfr. No. Description Diameter (Metric) For Use With (Application) No. of Positions Price Quantity & Availability  
Catalog Number Mfr. No. Description Diameter (Metric) For Use With (Application) No. of Positions Price Quantity & Availability  
88-882-149
View Documents
Thermo Scientific™
88882149
50 x 1.5/2.0 mL rotator drum 290 mm 1.5, 2.0 mL tubes 50
Each for $275.00
Add to cart
88-882-150
View Documents
Thermo Scientific™
88882150
30 x 15 mL rotator drum 290 mm 15 mL tubes 30
Each for $223.00
Add to cart
88-882-151
View Documents
Thermo Scientific™
88882151
15 x 50 mL rotator drum 290 mm 50 mL tubes 15
Each for $223.00
Add to cart
88-882-152
View Documents
Thermo Scientific™
88882152
10 x 10/15 mL and 20 x 5/7 mL rotator drum 290 mm 10/15 mL, 5/7 mL tubes 30
Each for $216.00
Add to cart
88-882-153
View Documents
Thermo Scientific™
88882153
64 x diameter 14 mm tube rotator drum 310 mm diameter 14 mm tube 64
Each for $223.00
Add to cart
88-882-154
View Documents
Thermo Scientific™
88882154
64 x diameter 18.5 mm tube rotator drum 310 mm diameter 18.5 mm tube 64
Each for $263.00
Add to cart
88-882-155
View Documents
Thermo Scientific™
88882155
142 x diameter 17 mm tube rotator drum 380 mm diameter 17 mm tube 142
Each for $433.00
Add to cart
88-882-156
View Documents
Thermo Scientific™
88882156
76 x diameter 26 mm tube rotator drum 380 mm diameter 26 mm tube 76
Each for $381.00
Add to cart
88-882-157
View Documents
Thermo Scientific™
88882157
60 x 30 diameter mm tube rotator drum 380 mm diameter 30 mm tube 60
Each for $381.00
Add to cart
Description
Description
Accessory for use with Digital Cel-Gro Tissue Culture Rotator, 120V and Digital Cel-Gro Tissue Culture Rotator, 230V
Specifications
Specifications
Rotating Drum