Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OHAUS™ Dry Block Heaters: PROMO Products for Science Education

Designed for applications that require repeatable results and superior temperature stability.

Specifications

Includes 92 in. Detachable, 3-wire cord and plug
Material Aluminum
Temperature Range (Metric) 5° above ambient to 100°C low range; 75°C to 150°C high range
View More Specs

Alert:

To avoid possible electrical hazard, do not fill well or block opening with water or other fluids. Unit is designed as a dry bath/incubator.


Products 4
Catalog Number Mfr. No. Type Electrical Requirements Heat Up Time Temperature Range (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Type Electrical Requirements Heat Up Time Temperature Range (Metric) Price Quantity & Availability  
S38781PMO Products for Science Education View Documents OHAUS™
30392045PMO
Dry Block Heater (1 block capacity) 45 min. 5° above ambient to 100°C low range; 75°C to 150°C high range This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
S38780PMO Products for Science Education View Documents OHAUS™
30392052PMO
Dry Block Heater (2 block capacity) 50 min. 5° above ambient to 100°C low range; 75°C to 150°C high range This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
S38782PMO Products for Science Education View Documents OHAUS™
30392080PMO
Dry Block Heater (2 block capacity) 120 V, 1.7 A, 210 w 50 min. 5° above ambient to 120°C This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
S38783PMO Products for Science Education View Documents OHAUS™
30392059PMO
Dry Block Heater (1 block capacity) 120 V, 0.9 A, 110 w 45 min. 5° above ambient to 120°C This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Dry block heaters require either one or two modular blocks, sold separately.

  • Ideal for incubation and activation of cultures, enzyme reactions, immunoassays, melting/boiling points and a wide variety of other laboratory procedures
  • Integral support rod holder with locking knob accepts optional external temperature probe kit
  • Accept separate interchangeable modular blocks (sold separately), accommodating various tube sizes from 0.2mL microtubes to 50mL centrifuge tubes
  • Each block has a thermometer well for measuring block temperature (blocks sold separately)
  • Efficient heater features close tube-and-block contact with a low-density design for exceptional temperature uniformity
  • Temperature range: ambient 5° to 120°C
  • With microprocessor control, LED display, audible alarm, temperature calibration mode

Applications:

Denaturing proteins, DNA applications, ELISA and other immunoassay studies

Order Info

Heaters require an Ohaus modular heating block for operation (sold separately). See S96607 for 1.5mL microcentrifuge tubes, S96608 for titer plates, S96609 for 15mL centrifuge tubes, S96610 for 50mL centrifuge tubes, S96611 for 10mm tubes, S96612 for 15/1

Warranty and Services

2-Year

Specifications

Specifications

92 in. Detachable, 3-wire cord and plug
Aluminum
5° above ambient to 100°C low range; 75°C to 150°C high range
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit