Learn More
Impacto™ DRYRIGGER CoolRigg Oil and Water Resistant Glove
Impacto™ DRYRIGGER COOLRIGG Gloves Help prevent hairline fractures, bruising and pinched fingers, Oil and Water Resistant protect the hand and fingers from impact and abrasion
Manufacturer:  Impacto™ WGCOOLRIGGXXL
 View more versions of this product
Catalog No. 19-400-084
Add to cart
Description
Description
Impacto™ Dryrigger CoolRigg Gloves
  • Mesh back for work in hot temperatures
  • Neoprene cuff offers excellent fit
  • High-visibility laminated fabric on the back and suede fabric on the palm repel oil and water
  • Under layer of PVC nitrile offers an extra barrier of protection in oily conditions
  • Silicone dots on the palm and fingertips provide excellent grip
  • Heavy duty TPR padded across the knuckles, back of fingers, thumb & back of the hand protects against bruises
Specifications
Specifications
11+ in.
Neoprene
Neoprene, Nitrile, PVC, Silicone, Suede
Oil, Water
3
Black, Gray, Yellow, Yellow, blue
Black
28+ cm
Full fingers
1
2X-Large
Powder-free
Non-sterile