Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DsbA-L Rabbit anti-Human, Mouse, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABS164425UL
Description
Anti-DsbA-L, Cat. No. ABS1644, is a rabbit polyclonal antibody that detects Glutathione S-transferase kappa 1 (DsbA-L) and is tested for use in Immunocytochemistry, Immunoprecipitation, and Western Blotting.
Specifications
DsbA-L | |
Polyclonal | |
Unconjugated | |
Glutathione S-transferase kappa 1;EC:2.5.1.18;GST 13-13;GST class-kappa;GSTK1-1;mGSTK1;Glutathione S-transferase subunit 13 | |
Rabbit | |
28 kDa observed. Uncharacterized bands may be observed in some lysate(s). | |
25 μL | |
Signaling | |
This rabbit polyclonal antibody detects DsbA-L (Glutathione S-transferase kappa 1) in human and mouse cells. | |
Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
IgG |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
Please refer to lot specific datasheet. | |
Rabbit polyclonal antiserum with 0.05% sodium azide with 20% glycerol. | |
Gstk1 | |
Myc-tagged full length recombinant protein from mouse. | |
Unpurified | |
RUO | |
Primary | |
Human, Mouse | |
Antibodies |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction