Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DSC1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549619
Description
Component of intercellular desmosome junctions. Involved in the interaction of plaque proteins and intermediate filaments mediating cell-cell adhesion. May contribute to epidermal cell positioning (stratification) by mediating differential adhesiveness between cells that express different isoforms. Linked to the keratinization of epithelial tissues.Specifications
DSC1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q08554 | |
DSC1 | |
KLH conjugated synthetic peptide between 659-687 amino acids from the C-terminal region of human DSC1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Western Blot | |
Unconjugated | |
DSC1 | |
Desmocollin-1, Cadherin family member 1, Desmosomal glycoprotein 2/3, DG2/DG3, DSC1, CDHF1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
1823 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction