Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dsDNA Mouse anti-Human, Clone: rDSD/4565, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen dsDNA
Clone rDSD/4565
Concentration 0.2 mg/ml
Dilution Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122813
View Documents
Novus Biologicals
NBP30767010UG
100 μg
Each for $374.34
Add to cart
 
NB122814
View Documents
Novus Biologicals
NBP30767020UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

dsDNA Monoclonal antibody specifically detects dsDNA in Human samples. It is validated for Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

dsDNA
0.2 mg/ml
Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
10 mM PBS with 0.05% BSA
IgG2a Kappa
Primary
Store at 4°C.
rDSD/4565
Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1-2 ug/ml
Unconjugated
Mouse
Human
Nuclei of Burkitt's cells
Monoclonal
Bioreactor concentrate
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit