Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DSDNA Mouse anti-Human, Clone: SPM603, Abnova™
Mouse monoclonal antibody raised against native human Double Stranded DNA (dsDNA).
Supplier: Abnova Corporation MAB13210
Specifications
DSDNA | |
Monoclonal | |
Unconjugated | |
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunocytochemistry (0.5-1 ug/mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1-2 ug/mL) The optimal working dilution should be d | |
Mouse | |
Protein G purification | |
RUO | |
Human | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (PFA fixed) | |
SPM603 | |
Mouse monoclonal antibody raised against native human Double Stranded DNA (dsDNA). | |
In 10mM PBS. | |
Nuclei of Burkitt's cells. | |
100 μg | |
Primary | |
Store at -20 to -80°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG3 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction