Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

dsDNA Rabbit anti-Human, Clone: DSD/4054R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30730210UG

 View more versions of this product

Catalog No. NB122077


Add to cart

Description

Description

dsDNA Monoclonal antibody specifically detects dsDNA in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

dsDNA
DSD/4054R
Unconjugated
10 mM PBS with 0.05% BSA
Rabbit
IgG
100 μg
Store at 4°C.
Monoclonal
Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
0.2 mg/ml
Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin
Unconjugated
Nuclei of Burkitt's cells
Bioreactor concentrate
RUO
Primary
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit