Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DSE Polyclonal Antibody
DSE Polyclonal Antibody
Supplier: Thermo Scientific PA565703
Description
DSE Polyclonal Antibody for ICC/IF

Specifications
DSE | |
Polyclonal | |
Unconjugated | |
DSE | |
6030499O08; AI480506; B130024B19Rik; Chondroitin-glucuronate 5-epimerase; dermatan sulfate epimerase; dermatan-sulfate epimerase; DS epimerase; Dse; DSEP; DSEPI; DS-epi1; EDSMC2; Sart2; SART-2; squamous cell carcinoma antigen recognized by T cells 2; squamous cell carcinoma antigen recognized by T-cells 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
29940 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UL01 | |
DSE | |
Recombinant Human DSE. Recombinant protein control fragment (Product #RP-106376). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction