Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ DSPG3 Polyclonal Antibody

Rabbit Polyclonal Antibody

$237.00 - $511.50

Specifications

Antigen DSPG3
Concentration 0.56 mg/mL
Content And Storage -20°C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5119095
View Documents
Invitrogen™
PA5119095
200 μL
Each for $511.50
Only null left
Add to Cart
 
PIPA5122025
View Documents
Invitrogen™
PA5122025
20 μL
Each for $237.00
Only null left
Add to Cart
 
Description

Description

DSPG3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications

Specifications

DSPG3
-20°C
Polyclonal
Liquid
IgG
Mouse
EPYC
Dermatan sulfate proteoglycan 3; dermatan sulphate proteoglycan 3; Dspg3; DSPG3proteoglycan-lb; Epiphycan; epiphycan proteoglycan; EPYC; Pglb; PG-Lb; Proteoglycan-Lb; SLRR3B; Small chondroitin/dermatan sulfate proteoglycan
EPYC
Primary
Antigen affinity chromatography
0.56 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300
P70186
13516
Recombinant protein Epiphycan. The antigen corresponds to amino acid range 130-322 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.