Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

DSPP Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP191612

 View more versions of this product

Catalog No. NBP191612



Description

Description

DSPP Polyclonal antibody specifically detects Antigen in Mouse samples. It is validated for Western Blotting.
Specifications

Specifications

DSPP
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
DSPP
Synthetic peptide directed towards the C terminal of mouse 1110051B16RIK. Peptide sequence PTTTPGEQGFHSQPPLQLLTQNRGHNPRESGGLAVPRLEDCTQVPAVNQH.
Immunogen affinity purified
RUO
Primary
1834
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Western Blot
Western Blot 1:1000
NP_899245
dentin phosphoprotein, dentin sialophosphoprotein, dentin sialoprotein, DFNA39, DMP3, DTDP2
Rabbit
IgG
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Expected identity based on immunogen sequence: Mouse: 100%.
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit