Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DsRed-Monomer Mouse anti-Tag, HRP, Clone: OTI4C7, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA183016

Description
DsRed-Monomer fluorescent protein.Specifications
DsRed-Monomer | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.1% BSA, 50% glycerol and 0.02% sodium azide | |
Mouse | |
Affinity Chromatography | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |
Western Blot | |
OTI4C7 | |
HRP | |
DsRed-Monomer | |
Recombit protein of DsRed-Monomer expressed in HEK293 cells. | |
100 μL | |
Primary | |
Antibody | |
IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction